Please login to your online account to display your discounted pricing

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  930001

 View more versions of this product

Catalog No. 02002848

  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 62.07
Chemical Name or Material Ethylene Glycol
CAS 107-21-1
Molecular Formula HOCH2CH2OH
Purity Grade BAKER ANALYZED™ Reagent
Flash Point 111.00°C
Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg

Specifications

Chemical Name or Material Ethylene Glycol
CAS 107-21-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HOCH2CH2OH
Formula Weight 62.07