Please login to your online account to display your discounted pricing

Ethylene Glycol AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ethylene Glycol
CAS 107-21-1
Molecular Formula HO(CH2)2OH
Formula Weight 62.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK50015

 
avantor performance
500119
Poly Pail 5gL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 62.07
Quantity 5gL
Packaging Poly Pail
Molecular Formula HO(CH2)2OH
Chemical Name or Material Ethylene Glycol
CAS 107-21-1