missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene glycol, 99+%, extra pure, ACROS Organics™

Manufacturer:  Acros Organics 146750010

 View more versions of this product

Catalog No. AC146750010



Specifications

Specifications

Ethylene glycol
107-21-1
Vapor Pressure: 0.12mmHg at 20°C
C2H6O2
MFCD00002885
01,375; 05,296; 09,217; 15,156
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
Viscous Liquid
Extra Pure
Authentic
1.4300 to 1.4330 (20°C, 589nm)
0.1% max. (Coulometric)
>500°C
Undesignated
1L
99+%
0.002% max.
99+%
HOCH2CH2OH
01, 465
15, 3852
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.06
≥99.0%
1.1130g/mL
Glass Bottle
0.05% max.
116°C (240.8°F)
196.0°C to 198.0°C
-13°C

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-473-3

RTECSNumber : KW2975000

TSCA : TSCA

SDS Documents