Please login to your online account to display your discounted pricing

Ethylene glycol, 99+%, extra pure, ACROS Organics™

$97.74 - $959.29

Chemical Identifiers

Chemical Name or Material Ethylene glycol
Name Note 99+%
CAS 107-21-1
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Formula Weight 62.06
MDL Number MFCD00002885
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146750010

  SDS
thermo fisher scientific
146750010
Glass Bottle 1L Each for $97.74

AC146750025

  SDS
fisher scientific acros
146750025
Glass Bottle 2.5L Each for $182.92

AC146750250

  SDS
thermo fisher scientific
146750250
Plastic Jerrycan 25L Each for $959.29
Description & Specifications

Specifications

Assay ≥99.0%
Melting Point −13°C
Physical Form Viscous Liquid
Purity Grade Extra Pure
Boiling Point 196.0° to 198.0°C
Formula Weight 62.06
Quantity 1L
Color Clear
Beilstein 01, 465
Merck Index 15, 3852
Molecular Formula C2H6O2
Packaging Glass Bottle
Flash Point 116°C (240.8°F)
TSCA TSCA
RTECS Number KW2975000
EINECS Number 203-473-3
Density 1.1130g/mL
Decomposition Information >500°C
Fieser 01,375; 05,296; 09,217; 15,156
Sulfated Ash 0.05% max.
Water 0.1% max. (Coulometric)
Synonym 1, 2-Ethanediol
Chemical Name or Material Ethylene glycol
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00002885
Refractive Index 1.4300 to 1.4330 (20°C, 589nm)
Infrared Spectrum Authentic
Additional Information Vapor Pressure: 0.12mmHg at 20°C
Acetic Acid 0.002% max.
Health Hazard 1 Warning
Linear Formula HOCH2CH2OH
CAS 107-21-1
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.