missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene glycol, 99.5%, for analysis, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 295530010
Description
For analysisSpecifications
Ethylene glycol | |
0.0002 meq/g max. | |
C2H6O2 | |
MFCD00002885 | |
01,375; 05,296; 09,217; 15,156 | |
1,2-dihydroxyethane, 1,2-ethanediol, 2-hydroxyethanol, ethylene alcohol, ethylene glycol, fridex, glycol, glycol alcohol, monoethylene glycol, tescol | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
Viscous Liquid | |
Analysis | |
1.1130g/mL | |
0.5ppm max. | |
1.4300 to 1.4330 | |
1.113 | |
0.1% max. (K.F.) | |
21 mPa.s (20°C) | |
Undesignated | |
1L |
107-21-1 | |
99.5% | |
HOCH2CH2OH | |
01, 465 | |
15, 3852 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.06 | |
99.5% | |
0.2ppm max. | |
Authentic | |
Glass bottle | |
Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons | |
0.005% max. | |
116°C | |
196.0°C to 198.0°C | |
-13.0°C |
Chemical Identifiers
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
MFCD00002885 | |
1,2-dihydroxyethane, 1,2-ethanediol, 2-hydroxyethanol, ethylene alcohol, ethylene glycol, fridex, glycol, glycol alcohol, monoethylene glycol, tescol | |
CHEBI:30742 | |
C(CO)O |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning