missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylene glycol, 99%

Manufacturer:  Alfa Aesar™ A1159136

 View more versions of this product

Catalog No. AAA1159136



Description

Description

Applications
Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

Solubility
Miscible with water, toluene, tetrahydrofuran, alcohol, chloroform and other organic solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents, strong bases, aldehydes and aluminum.
Specifications

Specifications

Ethylene glycol
C2H6O2
MFCD00002885
14,3798
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07
Sweet
196°C to 198°C
500g
107-21-1
HOCH2CH2OH
505945
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
99%
119°C (246°F)
-13°C

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
Safety and Handling

Safety and Handling

GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

GHS P Statement
P260-P264-P270-P301+P312-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KW2975000

TSCA : TSCA

SDS Documents