missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 410010025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethylene glycol | |
107-21-1 | |
196.0°C to 198.0°C | |
116°C | |
Glass Bottle | |
1.4300 to 1.4330 | |
1.113 | |
HOCH2CH2OH | |
01,375; 05,296; 09,217; 15,156 | |
Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
95% |
-13.0°C | |
1.1130g/mL | |
2.5 kg | |
Authentic | |
Viscous Liquid | |
95% | |
C2H6O2 | |
21 mPa.s (20°C) | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.06 | |
Pure |
Chemical Identifiers
107-21-1 | |
62.068 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
C2H6O2 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
Safety and Handling
Warning
RUO – Research Use Only