missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene Di(thiotosylate) [Protecting Reagent for Active Methylene], TCI America™

Manufacturer:  TCI America E047125G

 View more versions of this product

Catalog No. E047125G



Specifications

Specifications

Ethylene Di(thiotosylate) [Protecting Reagent for Active Methylene]
74°C
2225-23-2
MFCD00008550
DUFUGAKEFZRFEQ-UHFFFAOYSA-N
1-methyl-4-[2-(4-methylphenyl)sulfonylsulfanylethylsulfanylsulfonyl]benzene
75212
Crystal-Powder at 20°C
White
25g
C16H18O4S4
1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol Ditosylate, Ethylene Bis(p-toluenethiosulfonate), S,S′-Ethylene p-Toluenethiosulfonate
CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C2=CC=C(C=C2)C
402.556
402.56
≥98.0% (HPLC)

Chemical Identifiers

2225-23-2
402.556
DUFUGAKEFZRFEQ-UHFFFAOYSA-N
75212
CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C2=CC=C(C=C2)C
C16H18O4S4
MFCD00008550
1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol Ditosylate, Ethylene Bis(p-toluenethiosulfonate), S,S′-Ethylene p-Toluenethiosulfonate
1-methyl-4-[2-(4-methylphenyl)sulfonylsulfanylethylsulfanylsulfonyl]benzene
Safety and Handling

Safety and Handling

TSCA : No

Documents