Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylene 1,2-dithiocyanate, 98%

Chemical Identifiers

Chemical Name or Material Ethylene 1,2-dithiocyanate
CAS 629-17-4
Molecular Formula NCSCH2CH2SCN
Formula Weight 144.21
MDL Number MFCD00013807
UN Number UN2811
Synonym 1,2-Bis(thiocyanato)ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2218206

  SDS
alfa aesar™
B2218206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2218214

  SDS
alfa aesar™
B2218214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.21
Chemical Name or Material Ethylene 1,2-dithiocyanate
TSCA TSCA
Assay 98%
Molecular Formula NCSCH2CH2SCN
MDL Number MFCD00013807
CAS 629-17-4
UN Number UN2811
Synonym 1,2-Bis(thiocyanato)ethane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 88° to 92°C
Odor Irritating
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number XK9862500
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.