missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-piperazinecarboxylate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 118871000

 View more versions of this product

Catalog No. AC118871000



Description

Description

Specifications

Specifications

Ethyl N-piperazinecarboxylate
>110°C
Glass Bottle
1.08
273°C
100g
98.5% min. (GC)
MFCD00005964
1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-ethoxycarbonyl piperazine, 1-piperazinecarboxylic acid, ethyl ester, ethyl 1-piperazinecarboxylate, ethyl 1-piperazinocarboxylate, ethyl n-piperazinecarboxylate, ethylcarbonyl piperazine, n-carbethoxypiperazine, n-ethoxycarbonyl piperazine
LNOQURRKNJKKBU-UHFFFAOYSA-N
ethyl piperazine-1-carboxylate
8431
Oily Liquid
1.0800g/mL
Authentic
1.4760 to 1.478
0.1% max.
Yellow
120-43-4
C7H14N2O2
23, II, 9
Solubility in water: freely soluble
CCOC(=O)N1CCNCC1
158.2
158.2
99%

Chemical Identifiers

120-43-4
158.2
LNOQURRKNJKKBU-UHFFFAOYSA-N
8431
CCOC(=O)N1CCNCC1
C7H14N2O2
MFCD00005964
1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-ethoxycarbonyl piperazine, 1-piperazinecarboxylic acid, ethyl ester, ethyl 1-piperazinecarboxylate, ethyl 1-piperazinocarboxylate, ethyl n-piperazinecarboxylate, ethylcarbonyl piperazine, n-carbethoxypiperazine, n-ethoxycarbonyl piperazine
ethyl piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 204-395-2

SDS Documents