missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-(diphenylmethylene)glycinate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 124130050

 View more versions of this product

Catalog No. AC124130050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl N-(diphenylmethylene)glycinate
Authentic
White to Yellow
5g
97.5% min. (GC)
(C6H5)2C=NCH2COOCH2CH3
diphenylmethylene glycine ethyl ester, diphenylmethylene glycine ethylester, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, ethyl 2-diphenylmethylidene amino acetate, ethyl diphenylmethylenamino acetate, ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
267.32
267.32
98%
>110°C
Glass bottle
51°C to 55°C
69555-14-2
C17H17NO2
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl 2-(benzhydrylideneamino)acetate
319508
Crystalline Powder or Crystals

Chemical Identifiers

69555-14-2
267.32
QUGJYNGNUBHTNS-UHFFFAOYSA-N
319508
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
C17H17NO2
MFCD00010590
diphenylmethylene glycine ethyl ester, diphenylmethylene glycine ethylester, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, ethyl 2-diphenylmethylidene amino acetate, ethyl diphenylmethylenamino acetate, ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester
ethyl 2-(benzhydrylideneamino)acetate
SDS Documents
Product Certifications