missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl N-(diphenylmethylene)glycinate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC124130250
Specifications
Ethyl N-(diphenylmethylene)glycinate | |
Authentic | |
White to Yellow | |
25g | |
97.5% min. (GC) | |
(C6H5)2C=NCH2COOCH2CH3 | |
diphenylmethylene glycine ethyl ester, diphenylmethylene glycine ethylester, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, ethyl 2-diphenylmethylidene amino acetate, ethyl diphenylmethylenamino acetate, ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester | |
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 | |
267.32 | |
267.32 | |
98% |
>110°C | |
Glass bottle | |
51°C to 55°C | |
69555-14-2 | |
C17H17NO2 | |
MFCD00010590 | |
QUGJYNGNUBHTNS-UHFFFAOYSA-N | |
ethyl 2-(benzhydrylideneamino)acetate | |
319508 | |
Crystalline Powder or Crystals |
Chemical Identifiers
69555-14-2 | |
267.32 | |
QUGJYNGNUBHTNS-UHFFFAOYSA-N | |
319508 | |
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
C17H17NO2 | |
MFCD00010590 | |
diphenylmethylene glycine ethyl ester, diphenylmethylene glycine ethylester, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, ethyl 2-diphenylmethylidene amino acetate, ethyl diphenylmethylenamino acetate, ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester | |
ethyl 2-(benzhydrylideneamino)acetate |