Please login to your online account to display your discounted pricing

ethyleCrotonate, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  409865000

 View more versions of this product

Catalog No. AC409865000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Acid Value 1mg KOH/g max.
Density 0.918g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4237 to 1.4267 (20°C, 589nm)
Color Clear colorless
Quantity 500g
Chemical Name or Material ethylecrotonate
CAS 10544-63-5
Physical Form Clear colorless liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H10O2
Linear Formula CH3CH=CHCO2C2H5
Formula Weight 114.14g/mol
MDL Number MFCD00009289
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material ethylecrotonate
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C6H10O2
Linear Formula CH3CH=CHCO2C2H5
Formula Weight 114.14g/mol
MDL Number MFCD00009289
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents