Please login to your online account to display your discounted pricing

ethyleCrotonate, 98%, ACROS Organics™

$17.30 - $69.85

Chemical Identifiers

Chemical Name or Material ethylecrotonate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10O2
Linear Formula CH3CH=CHCO2C2H5
Formula Weight 114.14
MDL Number MFCD00009289
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC409865000

  SDS
thermo fisher scientific
AC409865000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC409860050

  SDS
thermo fisher scientific
409860050
Glass bottle 5g Each for $17.30

AC409861000

  SDS
thermo fisher scientific
409861000
Glass bottle 100g Each for $69.85
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 114.14
Quantity 500g
Color Clear colorless
Density 0.918g/mL
Acid Value 1mg KOH/g max.
Chemical Name or Material ethylecrotonate
Molecular Formula C6H10O2
MDL Number MFCD00009289
CAS 10544-63-5
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Refractive Index 1.4237 to 1.4267 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula CH3CH=CHCO2C2H5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.