missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyleCrotonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 409861000

 View more versions of this product

Catalog No. AC409861000


 

Specifications Description & Specifications

Specifications

≥97.5%
114.14
Clear colorless
ethylecrotonate
10544-63-5
Glass bottle
1mg KOH/g max.
98%
Liquid
100g
0.918g/mL
CH3CH=CHCO2C2H5
Authentic
1.4237 to 1.4267 (20°C, 589nm)
MFCD00009289
C6H10O2

Chemical Identifiers

98%
CH3CH=CHCO2C2H5
MFCD00009289
≥97.5%
C6H10O2
114.14
ethylecrotonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger