missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl chloroacetate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 118220010

 View more versions of this product

Catalog No. AC118220010



Description

Description

Specifications

Specifications

Ethyl chloroacetate
53°C
Glass bottle
1.15
142.0°C to 145.0°C
-26.0°C
105-39-5
C4H7ClO2
MFCD00000932
13,348
2-chloroacetic acid ethyl ester, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl alpha-chloroacetate, ethyl chloracetate, ethyl chloroacetate, ethyl chloroethanoate, ethyl monochloracetate, ethyl monochloroacetate, unii-6h07teh4tf
VEUUMBGHMNQHGO-UHFFFAOYSA-N
ethyl 2-chloroacetate
7751
Liquid
1.1500g/mL
Authentic
1.4200 to 1.4220
1.27 mPa.s (20°C)
Colorless
1L
98.5% min. (GC)
ClCH2CO2C2H5
02,197
15,3838
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
CCOC(=O)CCl
122.55
122.55
99%

Chemical Identifiers

105-39-5
122.55
VEUUMBGHMNQHGO-UHFFFAOYSA-N
7751
CCOC(=O)CCl
C4H7ClO2
MFCD00000932
2-chloroacetic acid ethyl ester, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl alpha-chloroacetate, ethyl chloracetate, ethyl chloroacetate, ethyl chloroethanoate, ethyl monochloracetate, ethyl monochloroacetate, unii-6h07teh4tf
ethyl 2-chloroacetate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Danger

EINECSNumber : 203-294-

SDS Documents