missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl carbazate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC118205000

 View more versions of this product

Catalog No. AC118205000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl carbazate
86°C
Glass bottle
White to Cream
500g
96% min. (GC)
H2NNHCO2C2H5
03, 98
carbethoxyhydrazine, carboethoxyhydrazine, ethoxycarbohydrazide, ethyl carbazate, ethyl carbazinate, ethyl hydrazinecarboxylate, ethylcarbazate, hydrazinecarboxylic acid, ethyl ester, monocarbethoxyhydrazine, n-carbethoxy hydrazine
CCOC(=O)NN
104.11
104.11
97%
86g/mL
Authentic
108°C to 110°C (22.0mmHg)
44°C to 48°C
4114-31-2
C3H8N2O2
MFCD00007595
01,360
VYSYZMNJHYOXGN-UHFFFAOYSA-N
ethyl N-aminocarbamate
20064
Powder

Chemical Identifiers

4114-31-2
104.11
VYSYZMNJHYOXGN-UHFFFAOYSA-N
20064
CCOC(=O)NN
C3H8N2O2
MFCD00007595
carbethoxyhydrazine, carboethoxyhydrazine, ethoxycarbohydrazide, ethyl carbazate, ethyl carbazinate, ethyl hydrazinecarboxylate, ethylcarbazate, hydrazinecarboxylic acid, ethyl ester, monocarbethoxyhydrazine, n-carbethoxy hydrazine
ethyl N-aminocarbamate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposur

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Get medical attention/advice if you feel unwell.
Do not eat,drink or smoke when using this pr

GHS Signal Word: Warning

EINECSNumber : 223-903-3

SDS Documents