missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl carbazate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 118200250
Specifications
Ethyl carbazate | |
86°C | |
Glass bottle | |
Cream to White | |
25g | |
96% min. (GC) | |
H2NNHCO2C2H5 | |
03, 98 | |
carbethoxyhydrazine, carboethoxyhydrazine, ethoxycarbohydrazide, ethyl carbazate, ethyl carbazinate, ethyl hydrazinecarboxylate, ethylcarbazate, hydrazinecarboxylic acid, ethyl ester, monocarbethoxyhydrazine, n-carbethoxy hydrazine | |
CCOC(=O)NN | |
104.11 | |
104.11 | |
97% |
86g/mL | |
Authentic | |
108°C to 110°C (22.0mmHg) | |
44°C to 48°C | |
4114-31-2 | |
C3H8N2O2 | |
MFCD00007595 | |
01,360 | |
VYSYZMNJHYOXGN-UHFFFAOYSA-N | |
ethyl N-aminocarbamate | |
20064 | |
Powder |
Chemical Identifiers
4114-31-2 | |
104.11 | |
VYSYZMNJHYOXGN-UHFFFAOYSA-N | |
20064 | |
CCOC(=O)NN |
C3H8N2O2 | |
MFCD00007595 | |
carbethoxyhydrazine, carboethoxyhydrazine, ethoxycarbohydrazide, ethyl carbazate, ethyl carbazinate, ethyl hydrazinecarboxylate, ethylcarbazate, hydrazinecarboxylic acid, ethyl ester, monocarbethoxyhydrazine, n-carbethoxy hydrazine | |
ethyl N-aminocarbamate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposur
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Get medical attention/advice if you feel unwell.
Do not eat,drink or smoke when using this pr
GHS Signal Word: Warning
EINECSNumber : 223-903-3