missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl benzoylacetate, 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 370851000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl benzoylacetate | |
Brown | |
265°C to 270°C | |
Authentic | |
Glass bottle | |
1.5240 to 1.531 | |
100 g | |
1.106 | |
ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate | |
GKKZMYDNDDMXSE-UHFFFAOYSA-N | |
ethyl 3-oxo-3-phenylpropanoate | |
7170 | |
90% |
94-02-0 | |
1.1060g/mL | |
140°C | |
88% min. (HPLC) | |
C11H12O3 | |
MFCD00009196 | |
10, 674 | |
15, 3822 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohols,ethers,miscible with oils | |
CCOC(=O)CC(=O)C1=CC=CC=C1 | |
192.21 | |
192.21 | |
Powder |
Chemical Identifiers
94-02-0 | |
192.21 | |
GKKZMYDNDDMXSE-UHFFFAOYSA-N | |
7170 | |
CCOC(=O)CC(=O)C1=CC=CC=C1 |
C11H12O3 | |
MFCD00009196 | |
ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate | |
ethyl 3-oxo-3-phenylpropanoate |
Safety and Handling
EINECSNumber : 202-295-3
RUO – Research Use Only