Please login to your online account to display your discounted pricing

ethyle7-hydroxy-4-oxo-4H-chromene-2-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10085EA

 View more versions of this product

Catalog No. BTB10085EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 234.21
Quantity 10g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
Chemical Name or Material Ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate
Molecular Formula C12H10O5
CAS 23866-72-0
CAS Min % 96.5
MDL Number MFCD00100382
Assay Percent Range 97%

Specifications

Chemical Name or Material Ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate
CAS 23866-72-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10O5
Formula Weight 234.21
MDL Number MFCD00100382