missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ethyle6-iodo-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB10203DA
Specifications
Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate | |
96.5 | |
97% | |
C12H9IO4 | |
1g | |
HUJFMOAXSPSGDU-UHFFFAOYSA-N | |
ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate | |
2775235 | |
97% |
35204-44-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD00100378 | |
ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester | |
CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1 | |
344.10 | |
344.1 |
Chemical Identifiers
35204-44-5 | |
344.10 | |
HUJFMOAXSPSGDU-UHFFFAOYSA-N | |
2775235 | |
CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1 |
C12H9IO4 | |
MFCD00100378 | |
ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester | |
ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate |