Please login to your online account to display your discounted pricing

ethyle6-iodo-4-oxo-4H-chromene-2-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10203EA

 View more versions of this product

Catalog No. BTB10203EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 344.1
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H9IO4
MDL Number MFCD00100378
CAS 35204-44-5
CAS Min % 96.5

Specifications

Chemical Name or Material Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
CAS 35204-44-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H9IO4
Formula Weight 344.1
MDL Number MFCD00100378