Please login to your online account to display your discounted pricing

ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate
CAS 64067-99-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8ClN3O2
Formula Weight 225.63
MDL Number MFCD03086230
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00216CB

  SDS
fisher scientific company
MO00216CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.63
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate
Molecular Formula C9H8ClN3O2
MDL Number MFCD03086230
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 64067-99-8