Please login to your online account to display your discounted pricing

ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00216CB

 View more versions of this product

Catalog No. MO00216CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 225.63
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8ClN3O2
MDL Number MFCD03086230
CAS 64067-99-8

Specifications

Chemical Name or Material Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate
CAS 64067-99-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8ClN3O2
Formula Weight 225.63
MDL Number MFCD03086230