Please login to your online account to display your discounted pricing

ethyle5-pyrimidinecarboxylate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ethyle5-pyrimidinecarboxylate
Assay Percent Range 97%
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD03094043
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC425940050

  SDS
thermo fisher scientific
AC425940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC425940010

  SDS
thermo fisher scientific
AC425940010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.15
Quantity 5g
Density 1.16g/mL
CAS 40929-50-8
Assay Percent Range 97%
Chemical Name or Material ethyle5-pyrimidinecarboxylate
Molecular Formula C7H8N2O2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD03094043
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.