Learn More
Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 349730010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 5-(chloromethyl)-2-furancarboxylate | |
1.2250g/mL | |
>110°C | |
94% min. (GC) | |
C8H9ClO3 | |
MFCD00173837 | |
1.225 | |
JBACYJRMCXLIQU-UHFFFAOYSA-N | |
ethyl 5-(chloromethyl)furan-2-carboxylate | |
263166 | |
95% |
2528-00-9 | |
115.0°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.5130 | |
1g | |
ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate | |
CCOC(=O)C1=CC=C(O1)CCl | |
188.61 | |
188.61 |
Chemical Identifiers
2528-00-9 | |
188.61 | |
JBACYJRMCXLIQU-UHFFFAOYSA-N | |
263166 | |
CCOC(=O)C1=CC=C(O1)CCl |
C8H9ClO3 | |
MFCD00173837 | |
ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate | |
ethyl 5-(chloromethyl)furan-2-carboxylate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only