missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-aminobenzo[b]furan-2-carboxylate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437970050

 View more versions of this product

Catalog No. AC437970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 5-aminobenzo[b]furan-2-carboxylate
Beige
5g
96% min. (GC)
2-benzofurancarboxylic acid, 5-amino-, ethyl ester, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-amino-benzofuran-2-carboxylic acid ethyl ester, 5-aminobenzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, ethyl 5-aminobenzo d furan-2-carboxylate, ethyl 5-aminobenzofuran-2-carboxylate, ethylaminobenzofurancarboxylate, ksc494o5f, pubchem23808
CCOC(=O)C1=CC2=C(O1)C=CC(=C2)N
205.22
205.22
97%
Authentic
56°C to 64°C
174775-48-5
C11H11NO3
YFFLLDHEEWSHQG-UHFFFAOYSA-N
ethyl 5-amino-1-benzofuran-2-carboxylate
9837138
Powder

Chemical Identifiers

174775-48-5
205.22
2-benzofurancarboxylic acid, 5-amino-, ethyl ester, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-amino-benzofuran-2-carboxylic acid ethyl ester, 5-aminobenzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, ethyl 5-aminobenzo d furan-2-carboxylate, ethyl 5-aminobenzofuran-2-carboxylate, ethylaminobenzofurancarboxylate, ksc494o5f, pubchem23808
ethyl 5-amino-1-benzofuran-2-carboxylate
C11H11NO3
YFFLLDHEEWSHQG-UHFFFAOYSA-N
9837138
CCOC(=O)C1=CC2=C(O1)C=CC(=C2)N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS Documents