Please login to your online account to display your discounted pricing

ethyle5-amino-1-methylpyrazole-4-carboxylate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 5-amino-1-methylpyrazole-4-carboxylate
CAS 31037-02-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H11N3O2
Formula Weight 169.18
MDL Number MFCD00051652
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC355180050

  SDS
thermo fisher scientific
AC355180050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC355180010

  SDS
thermo fisher scientific
35518-0010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 169.18
Quantity 5g
Color Yellow
Melting Point 98° to 102°C
Assay Percent Range 98%
Chemical Name or Material Ethyl 5-amino-1-methylpyrazole-4-carboxylate
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00051652
Packaging Glass bottle
Molecular Formula C7H11N3O2
Infrared Spectrum Authentic
CAS 31037-02-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.