Please login to your online account to display your discounted pricing

ethyle5-amino-1-methylpyrazole-4-carboxylate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35518-0010

 View more versions of this product

Catalog No. AC355180010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 169.18
Quantity 1g
Color Yellow
Melting Point 98° to 102°C
Chemical Name or Material Ethyl 5-amino-1-methylpyrazole-4-carboxylate
Assay Percent Range 98%
Molecular Formula C7H11N3O2
CAS 31037-02-2
Packaging Glass bottle
MDL Number MFCD00051652
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Ethyl 5-amino-1-methylpyrazole-4-carboxylate
CAS 31037-02-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H11N3O2
Formula Weight 169.18
MDL Number MFCD00051652
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents