Please login to your online account to display your discounted pricing

ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC23210DE

 View more versions of this product

Catalog No. CC23210DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 302.15
Quantity 5g
Assay 97%
CAS Max % 100.0
CAS 423768-50-7
Assay Percent Range 97%
Chemical Name or Material ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate
CAS Min % 96.5
MDL Number MFCD03407336
Molecular Formula C10H8BrNO3S

Specifications

Chemical Name or Material ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate
CAS 423768-50-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8BrNO3S
Formula Weight 302.15
MDL Number MFCD03407336