missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle4-pyridylacetate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC425930010

 View more versions of this product

Catalog No. AC425930010


 

Specifications Description & Specifications

Specifications

165.19
1.079g/mL
97%
54401-85-3
Glass bottle
1g
ethyle4-pyridylacetate
C9H11NO2
MFCD00051830

Chemical Identifiers

97%
165.19
ethyle4-pyridylacetate
C9H11NO2
MFCD00051830

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.