Please login to your online account to display your discounted pricing

ethyle4-pyridylacetate, 97%, ACROS Organics™

$34.22 - $34.22

Chemical Identifiers

Chemical Name or Material ethyle4-pyridylacetate
Assay Percent Range 97%
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00051830
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC425930010

 
thermo fisher scientific
AC425930010
Glass bottle 1g Each for $34.22

AC425930050

 
thermo fisher scientific
AC425930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.19
Quantity 1g
Density 1.079g/mL
Chemical Name or Material ethyle4-pyridylacetate
Assay Percent Range 97%
Molecular Formula C9H11NO2
MDL Number MFCD00051830
CAS 54401-85-3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.