missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle4-nitrocinnamate, 99%, Predominantly trans, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 160740250

 View more versions of this product

Catalog No. AC160740250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
221.21
Light yellow-beige to yellow
09, 607
Authentic
O2NC6H4CH=CHCO2C2H5
MFCD00007380
Predominantly trans
Fine Crystalline Powder
25g
137° to 140°C
Glass bottle
C11H11NO4
99%
Ethyl 4-nitrocinnamate
953-26-4

Chemical Identifiers

953-26-4
C11H11NO4
221.21
Ethyl 4-nitrocinnamate, Predominantly trans
99%
O2NC6H4CH=CHCO2C2H5
MFCD00007380
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 213-463-0

RTECSNumber : GE0353332

TSCA : TSCA