Please login to your online account to display your discounted pricing

ethyle4-nitrocinnamate, 99%, Predominantly trans, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 4-nitrocinnamate
Name Note Predominantly trans
CAS 953-26-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H11NO4
Linear Formula O2NC6H4CH=CHCO2C2H5
Formula Weight 221.21
MDL Number MFCD00007380
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160740250

  SDS
thermo fisher scientific
160740250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 221.21
Quantity 25g
Color Light yellow-beige to yellow
EINECS Number 213-463-0
TSCA TSCA
RTECS Number GE0353332
Melting Point 137° to 140°C
Beilstein 09, 607
Chemical Name or Material Ethyl 4-nitrocinnamate
Name Note Predominantly trans
Assay Percent Range 99%
Molecular Formula C11H11NO4
Linear Formula O2NC6H4CH=CHCO2C2H5
MDL Number MFCD00007380
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 953-26-4