Please login to your online account to display your discounted pricing

ethyle4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 4-methyl-2-phenyl-1, 3-thiazole-5-carboxylate
CAS 53715-64-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H13NO2S
Formula Weight 247.32
MDL Number MFCD00141953
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB02157EA

 
fisher scientific company
SPB02157EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.32
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 4-methyl-2-phenyl-1, 3-thiazole-5-carboxylate
Packaging Amber Glass Bottle
Molecular Formula C13H13NO2S
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
MDL Number MFCD00141953
CAS 53715-64-3
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.