Please login to your online account to display your discounted pricing

ethyle4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylate
CAS 175277-03-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H12F3NO2S
Formula Weight 315.31
MDL Number MFCD00068104
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB03021EA

 
fisher scientific company
SPB03021EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 315.31
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylate
Molecular Formula C14H12F3NO2S
CAS 175277-03-9
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00068104