Please login to your online account to display your discounted pricing

ethyle4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB03021EA

 View more versions of this product

Catalog No. SPB03021EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 315.31
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylate
Molecular Formula C14H12F3NO2S
CAS 175277-03-9
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00068104

Specifications

Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylate
CAS 175277-03-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H12F3NO2S
Formula Weight 315.31
MDL Number MFCD00068104