Please login to your online account to display your discounted pricing

ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
CAS 175276-52-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H17NO3
Formula Weight 271.31
MDL Number MFCD00204225
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00028DA

 
fisher scientific company
SP00028DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.31
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C16H17NO3
MDL Number MFCD00204225
CAS 175276-52-5