Please login to your online account to display your discounted pricing

ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SP00028DA

 View more versions of this product

Catalog No. SP00028DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 271.31
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C16H17NO3
MDL Number MFCD00204225
CAS 175276-52-5

Specifications

Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
CAS 175276-52-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H17NO3
Formula Weight 271.31
MDL Number MFCD00204225