Learn More
Ethyl 4-dimethylaminobenzoate, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC118405000
Specifications
Ethyl 4-dimethylaminobenzoate | |
Authentic | |
1.06 | |
62°C to 65°C | |
10287-53-3 | |
C11H15NO2 | |
MFCD00009115 | |
benzoic acid, 4-dimethylamino-, ethyl ester, ethyl 4-dimethylamino benzoate, ethyl p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, ethyl-p-dimethylaminobenzoate, kayacure epa, n,n-dimethylbenzocaine, parbenate, speedcure edb, unii-829s8d3y0x | |
FZUGPQWGEGAKET-UHFFFAOYSA-N | |
ethyl 4-(dimethylamino)benzoate | |
25127 | |
193.25 | |
99+% |
1.0600g/mL | |
Plastic bottle | |
White to Yellow | |
500g | |
99% min. (GC) | |
(CH3)2NC6H4CO2C2H5 | |
14, I, 571 | |
Solubility in water: insoluble. Other solubilities: >50g/100 mL acetone (20 c),18g/100mL ethanol (20 c),2g/100mL mineral oil (20 c) | |
CCOC(=O)C1=CC=C(C=C1)N(C)C | |
193.25 | |
CHEBI:52073 | |
Crystalline Powder |
Chemical Identifiers
10287-53-3 | |
193.25 | |
FZUGPQWGEGAKET-UHFFFAOYSA-N | |
25127 | |
ethyl 4-(dimethylamino)benzoate |
C11H15NO2 | |
MFCD00009115 | |
benzoic acid, 4-dimethylamino-, ethyl ester, ethyl 4-dimethylamino benzoate, ethyl p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, ethyl-p-dimethylaminobenzoate, kayacure epa, n,n-dimethylbenzocaine, parbenate, speedcure edb, unii-829s8d3y0x | |
CHEBI:52073 | |
CCOC(=O)C1=CC=C(C=C1)N(C)C |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 233-634-3