Please login to your online account to display your discounted pricing

ethyle4-chlorobutyrate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ethyle4-chlorobutyrate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11ClO2
Linear Formula Cl(CH2)3CO2C2H5
Formula Weight 150.6
MDL Number MFCD00001004
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118241000

  SDS
thermo fisher scientific
118241000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC118240050

  SDS
thermo fisher scientific
118240050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC118245000

  SDS
thermo fisher scientific
AC118245000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Color Clear yellow to brownish
Formula Weight 150.6
Quantity 100g
Beilstein 02, 278
Density 1.07g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4296 to 1.4316 (20°C, 589nm)
Molecular Formula C6H11ClO2
MDL Number MFCD00001004
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material ethyle4-chlorobutyrate
Assay Percent Range 97%
Linear Formula Cl(CH2)3CO2C2H5
CAS 3153-36-4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.