missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163750050

 View more versions of this product

Catalog No. AC163750050



Specifications

Specifications

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate
Glass bottle
60.0°C to 63.0°C
5909-24-0
C8H9ClN2O2S
2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)SC
232.68
232.68
98%
Authentic
Beige to White
5g
97.5% min. (GC)
MFCD00006085
SNNHLSHDDGJVDM-UHFFFAOYSA-N
ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate
80008
Crystalline Powder

Chemical Identifiers

5909-24-0
232.68
SNNHLSHDDGJVDM-UHFFFAOYSA-N
80008
CCOC(=O)C1=CN=C(N=C1Cl)SC
C8H9ClN2O2S
MFCD00006085
2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate
ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 227-619-

SDS Documents