Please login to your online account to display your discounted pricing

ethyle4-acetylbenzoate, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397310010

 View more versions of this product

Catalog No. AC397310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 192.21
Physical Form Crystalline Powder
Quantity 1g
Color White to beige
Melting Point 54° to 58°C
Packaging Glass bottle
Linear Formula CH3CH2OOCC6H4COCH3
Molecular Formula C11H12O3
Chemical Name or Material ethyle4-acetylbenzoate
CAS 38430-55-6
MDL Number MFCD00013241
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material ethyle4-acetylbenzoate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12O3
Linear Formula CH3CH2OOCC6H4COCH3
Formula Weight 192.21
MDL Number MFCD00013241
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents