Please login to your online account to display your discounted pricing

ethyle4-acetylbenzoate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ethyle4-acetylbenzoate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12O3
Linear Formula CH3CH2OOCC6H4COCH3
Formula Weight 192.21
MDL Number MFCD00013241
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC397310050

 
thermo fisher scientific
AC397310050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC397310010

 
thermo fisher scientific
AC397310010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 192.21
Quantity 5g
Color White to beige
Melting Point 54° to 58°C
Packaging Glass bottle
CAS 38430-55-6
Infrared Spectrum Authentic
Chemical Name or Material ethyle4-acetylbenzoate
Assay Percent Range 99%
Molecular Formula C11H12O3
MDL Number MFCD00013241
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Linear Formula CH3CH2OOCC6H4COCH3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.