Please login to your online account to display your discounted pricing

ethyle4,6-dichloro-2-(methylthio)quinoline-3-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material ethyle4, 6-dichloro-2-(methylthio)quinoline-3-carboxylate
CAS 227958-96-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11Cl2NO2S
Formula Weight 316.21
MDL Number MFCD00276616
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM08739DA

  SDS
fisher scientific company
KM08739DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM08739EA

  SDS
fisher scientific company
KM08739EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 316.21
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material ethyle4, 6-dichloro-2-(methylthio)quinoline-3-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C13H11Cl2NO2S
MDL Number MFCD00276616
CAS 227958-96-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.