Please login to your online account to display your discounted pricing

ethyle4,6-dichloro-2-(methylthio)quinoline-3-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM08739EA

 View more versions of this product

Catalog No. KM08739EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 316.21
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material ethyle4, 6-dichloro-2-(methylthio)quinoline-3-carboxylate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00276616
Packaging Amber Glass Bottle
CAS 227958-96-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 97%
Molecular Formula C13H11Cl2NO2S

Specifications

Chemical Name or Material ethyle4, 6-dichloro-2-(methylthio)quinoline-3-carboxylate
CAS 227958-96-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H11Cl2NO2S
Formula Weight 316.21
MDL Number MFCD00276616
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents