Please login to your online account to display your discounted pricing

ethyle4,4,4-trifluorocrotonate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC307360050

 View more versions of this product

Catalog No. AC307360050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 168.12
Quantity 5g
Color Clear colorless
Density 1.1200g/mL
Chemical Name or Material Ethyl 4, 4, 4-trifluorocrotonate
Packaging Glass bottle
Molecular Formula C6H7F3O2
Assay Percent Range 98%
CAS 25597-16-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.3591 to 1.3611 (20°C, 589nm)
Linear Formula CF3CH=CHCO2C2H5
MDL Number MFCD00009903
Synonym Ethyl 4, 4, 4-trifluoro-trans-2-butenoate
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Ethyl 4, 4, 4-trifluorocrotonate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7F3O2
Linear Formula CF3CH=CHCO2C2H5
Formula Weight 168.12
MDL Number MFCD00009903
Synonym Ethyl 4, 4, 4-trifluoro-trans-2-butenoate
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents