Please login to your online account to display your discounted pricing

ethyle4,4,4-trifluorocrotonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 4, 4, 4-trifluorocrotonate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7F3O2
Linear Formula CF3CH=CHCO2C2H5
Formula Weight 168.12
MDL Number MFCD00009903
Synonym Ethyl 4, 4, 4-trifluoro-trans-2-butenoate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC307360050

  SDS
thermo fisher scientific
AC307360050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC307360010

  SDS
thermo fisher scientific
307360010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 168.12
Physical Form Liquid
Quantity 5g
Color Clear colorless
Density 1.1200g/mL
Packaging Glass bottle
Chemical Name or Material Ethyl 4, 4, 4-trifluorocrotonate
Molecular Formula C6H7F3O2
CAS 25597-16-4
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.3591 to 1.3611 (20°C, 589nm)
MDL Number MFCD00009903
Synonym Ethyl 4, 4, 4-trifluoro-trans-2-butenoate
Linear Formula CF3CH=CHCO2C2H5
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.