missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle4,4,4-trifluorocrotonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 307360010

 View more versions of this product

Catalog No. AC307360010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
168.12
Clear colorless
CF3CH=CHCO2C2H5
25597-16-4
C6H7F3O2
Glass bottle
1.3591 to 1.3611 (20°C, 589nm)
Liquid
1g
1.1200g/mL
Ethyl 4, 4, 4-trifluorocrotonate
98%
MFCD00009903
Authentic
Ethyl 4, 4, 4-trifluoro-trans-2-butenoate

Chemical Identifiers

98%
CF3CH=CHCO2C2H5
MFCD00009903
Ethyl 4, 4, 4-trifluorocrotonate
C6H7F3O2
168.12
Ethyl 4, 4, 4-trifluoro-trans-2-butenoate
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning