missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-isocyanatopropionate, 98%, Acros Organics™

Chemical Identifiers

CAS 5100-34-5
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD00078461
InChI Key XBSGYVHOINMTIM-UHFFFAOYSA-N
Synonym ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
PubChem CID 2733373
IUPAC Name ethyl 3-isocyanatopropanoate
SMILES CCOC(=O)CCN=C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC319270010 SDS View Documents Acros Organics
319270010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

5100-34-5
143.142
XBSGYVHOINMTIM-UHFFFAOYSA-N
2733373
CCOC(=O)CCN=C=O
C6H9NO3
MFCD00078461
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
ethyl 3-isocyanatopropanoate

Specifications

1.0870g/mL
Glass bottle
1.087
Colorless to Yellow
5100-34-5
C6H9NO3
MFCD00078461
XBSGYVHOINMTIM-UHFFFAOYSA-N
ethyl 3-isocyanatopropanoate
2733373
Liquid
Ethyl 3-isocyanatopropionate
Authentic
1.4275 to 1.4295
68.0°C (2.0 mmHg)
1g
98%
OCNCH2CH2C(=O)OCH2CH3
ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
CCOC(=O)CCN=C=O
143.142
143.07
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning